Product Details

SNP ID
rs200157397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48059016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCAGTATGAAGTCTTTGATGGC[C/T]AGTGAGGTGTGACTTCTGGGAAAAG
Phenotype
MIM: 300819
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF630 PubMed Links

Gene Details

Gene
ZNF630
Gene Name
zinc finger protein 630
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037735.3 1989 Missense Mutation AGC,GGC S476G NP_001032824.2
NM_001190255.2 1989 Missense Mutation AGC,GGC S462G NP_001177184.1
NM_001282201.1 1989 Missense Mutation AGC,GGC S476G NP_001269130.1
NM_001282202.1 1989 Missense Mutation AGC,GGC S352G NP_001269131.1
Gene
ZNF630-AS1
Gene Name
ZNF630 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details