Product Details

SNP ID
rs201428513
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:76176889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTTTTGCCTTTATAGAAGTGGAG[A/G]CCATTCTGCTTGAAGTTTAATGGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PBDC1 PubMed Links

Gene Details

Gene
PBDC1
Gene Name
polysaccharide biosynthesis domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300888.1 509 Intron NP_001287817.1
NM_016500.3 509 Silent Mutation AGA,AGG R102R NP_057584.2

View Full Product Details