Product Details

SNP ID
rs200833768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:78131683 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGCTTCATTGGCTGTGTTCTGT[C/G]ACGAAACCATGTTGGTGGTAATAAG
Phenotype
MIM: 311800 MIM: 300754
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PGK1 PubMed Links

Gene Details

Gene
PGK1
Gene Name
phosphoglycerate kinase 1
There are no transcripts associated with this gene.

Gene
TAF9B
Gene Name
TATA-box binding protein associated factor 9b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015975.4 Intron NP_057059.2

View Full Product Details