Product Details

SNP ID
rs200840695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:76175556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTGAGTTCCGGAAAAATTTTGA[A/G]ACCCTTAGGATAGATGTGTTGGACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PBDC1 PubMed Links

Gene Details

Gene
PBDC1
Gene Name
polysaccharide biosynthesis domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300888.1 443 Silent Mutation GAA,GAG E80E NP_001287817.1
NM_016500.3 443 Silent Mutation GAA,GAG E80E NP_057584.2

View Full Product Details