Product Details
- SNP ID
-
rs9444105
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:83869940 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGCCCCCAAGAATAGTGCTGAAGT[G/T]CTGTGGCAAGAAAGCTGTAGTATGC
- Phenotype
-
MIM: 608343
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CYB5R4
PubMed Links
Gene Details
- Gene
- CYB5R4
- Gene Name
- cytochrome b5 reductase 4
There are no transcripts associated with this gene.
View Full Product Details