Product Details

SNP ID
rs12851
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:30005656 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCCCGGCCCCCTCCAGGGGACAGTT[G/A]TTTAAACGAGTGGCCGGGAGCATCT
Phenotype
MIM: 604567 MIM: 603365
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
DOC2A PubMed Links

Gene Details

Gene
DOC2A
Gene Name
double C2 domain alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282062.1 1925 UTR 3 NP_001268991.1
NM_001282063.1 1925 UTR 3 NP_001268992.1
NM_001282068.1 1925 UTR 3 NP_001268997.1
NM_003586.2 1925 UTR 3 NP_003577.2
XM_011545975.1 1925 UTR 3 XP_011544277.1
XM_017023776.1 1925 UTR 3 XP_016879265.1
XM_017023777.1 1925 UTR 3 XP_016879266.1
XM_017023778.1 1925 UTR 3 XP_016879267.1
Gene
HIRIP3
Gene Name
HIRA interacting protein 3
There are no transcripts associated with this gene.

Gene
INO80E
Gene Name
INO80 complex subunit E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304562.1 1925 UTR 3 NP_001291491.1
NM_001304563.1 1925 UTR 3 NP_001291492.1
NM_173618.2 1925 UTR 3 NP_775889.1
XM_011545809.2 1925 UTR 3 XP_011544111.1
XM_011545811.2 1925 Intron XP_011544113.1
XM_011545812.2 1925 Intron XP_011544114.1
XM_017023169.1 1925 Intron XP_016878658.1

View Full Product Details