Product Details

SNP ID
rs11977237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:87336025 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTCTAGGTTTCACTTACCACAA[A/C]TATGATACAACTCATTGATTTCATA
Phenotype
MIM: 606090 MIM: 616403
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CROT PubMed Links

Gene Details

Gene
CROT
Gene Name
carnitine O-octanoyltransferase
There are no transcripts associated with this gene.

Gene
TP53TG1
Gene Name
TP53 target 1 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details