Product Details

SNP ID
rs1919313
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:18090695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTTCACTCCAGAGGCAGCCAGTA[A/G]TGGTCAGAGATCATATTCTGGGTGT
Phenotype
MIM: 606543
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HDAC9 PubMed Links

Gene Details

Gene
HDAC9
Gene Name
histone deacetylase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204144.2 Intron NP_001191073.1
NM_001204145.2 Intron NP_001191074.1
NM_001204146.2 Intron NP_001191075.1
NM_001204147.2 Intron NP_001191076.1
NM_001204148.2 Intron NP_001191077.1
NM_001321868.1 Intron NP_001308797.1
NM_001321869.1 Intron NP_001308798.1
NM_001321870.1 Intron NP_001308799.1
NM_001321871.1 Intron NP_001308800.1
NM_001321872.1 Intron NP_001308801.1
NM_001321873.1 Intron NP_001308802.1
NM_001321874.1 Intron NP_001308803.1
NM_001321875.1 Intron NP_001308804.1
NM_001321876.1 Intron NP_001308805.1
NM_001321877.1 Intron NP_001308806.1
NM_001321878.1 Intron NP_001308807.1
NM_001321879.1 Intron NP_001308808.1
NM_001321884.1 Intron NP_001308813.1
NM_001321885.1 Intron NP_001308814.1
NM_001321886.1 Intron NP_001308815.1
NM_001321887.1 Intron NP_001308816.1
NM_001321888.1 Intron NP_001308817.1
NM_001321889.1 Intron NP_001308818.1
NM_001321890.1 Intron NP_001308819.1
NM_001321891.1 Intron NP_001308820.1
NM_001321893.1 Intron NP_001308822.1
NM_001321894.1 Intron NP_001308823.1
NM_001321895.1 Intron NP_001308824.1
NM_001321896.1 Intron NP_001308825.1
NM_001321897.1 Intron NP_001308826.1
NM_001321898.1 Intron NP_001308827.1
NM_001321899.1 Intron NP_001308828.1
NM_001321900.1 Intron NP_001308829.1
NM_001321901.1 Intron NP_001308830.1
NM_001321902.1 Intron NP_001308831.1
NM_014707.3 Intron NP_055522.1
NM_058176.2 Intron NP_478056.1
NM_178423.2 Intron NP_848510.1
NM_178425.3 Intron NP_848512.1
XM_011515629.2 Intron XP_011513931.1
XM_011515631.2 Intron XP_011513933.1
XM_011515634.2 Intron XP_011513936.1
XM_011515635.2 Intron XP_011513937.1
XM_011515636.2 Intron XP_011513938.1
XM_011515637.2 Intron XP_011513939.1
XM_011515638.2 Intron XP_011513940.1
XM_011515639.2 Intron XP_011513941.1
XM_011515641.2 Intron XP_011513943.1
XM_011515642.2 Intron XP_011513944.1
XM_011515644.2 Intron XP_011513946.1
XM_011515645.2 Intron XP_011513947.1
XM_011515646.2 Intron XP_011513948.1
XM_011515648.2 Intron XP_011513950.1
XM_011515649.2 Intron XP_011513951.1
XM_011515651.1 Intron XP_011513953.1
XM_017012826.1 Intron XP_016868315.1
XM_017012827.1 Intron XP_016868316.1
XM_017012828.1 Intron XP_016868317.1
XM_017012829.1 Intron XP_016868318.1
XM_017012830.1 Intron XP_016868319.1
XM_017012831.1 Intron XP_016868320.1
XM_017012832.1 Intron XP_016868321.1

View Full Product Details