Product Details

SNP ID
rs11385
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65719127 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCAAGCGGCTCCTGCGGATCGA[C/A]TCCAAGTGTCTGCACTTCACTCCCA
Phenotype
MIM: 601409 MIM: 610330
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
KAT5 PubMed Links

Gene Details

Gene
KAT5
Gene Name
lysine acetyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206833.1 1331 Silent Mutation GAA,GAC E477D NP_001193762.1
NM_006388.3 1331 Silent Mutation GAA,GAC E496D NP_006379.2
NM_182709.2 1331 Silent Mutation GAA,GAC E444D NP_874368.1
NM_182710.2 1331 Silent Mutation GAA,GAC E529D NP_874369.1
XM_006718421.2 1331 Missense Mutation GAA,GAC E505D XP_006718484.1
Gene
RNASEH2C
Gene Name
ribonuclease H2 subunit C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032193.3 1331 UTR 3 NP_115569.2

View Full Product Details