Product Details

SNP ID
rs61757484
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:6425489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGCAGCGAGTGCCCTGGGGACG[A/G]GGGTGGCTCTGTGGCTGGCTCTGCA
Phenotype
MIM: 604392
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AIPL1 PubMed Links

Gene Details

Gene
AIPL1
Gene Name
aryl hydrocarbon receptor interacting protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033054.2 1149 Missense Mutation CCG,TCG P313S NP_001028226.1
NM_001033055.2 1149 Missense Mutation CCG,TCG P316S NP_001028227.1
NM_001285399.2 1149 Missense Mutation CCG,TCG P364S NP_001272328.1
NM_001285400.2 1149 Missense Mutation CCG,TCG P354S NP_001272329.1
NM_001285401.2 1149 Missense Mutation CCG,TCG P352S NP_001272330.1
NM_001285402.1 1149 Missense Mutation CCG,TCG P337S NP_001272331.1
NM_001285403.2 1149 Intron NP_001272332.1
NM_014336.4 1149 Missense Mutation CCG,TCG P376S NP_055151.3

View Full Product Details