Product Details

SNP ID
rs1872900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143429825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGTGGTGGTGGGCGGCGTGGTG[A/G]TGGTGGGCGGCGTGGTGCGCGCCGT
Phenotype
MIM: 610303
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAFA PubMed Links

Gene Details

Gene
MAFA
Gene Name
MAF bZIP transcription factor A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_201589.3 608 Silent Mutation CAC,CAT H194H NP_963883.2
Gene
ZC3H3
Gene Name
zinc finger CCCH-type containing 3
There are no transcripts associated with this gene.

View Full Product Details