Product Details

SNP ID
rs120074114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44948767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCGACCTTCCTCACCCAGGTGA[A/C]GGAATCTCTCTCCAGTTACTGGGAG
Phenotype
MIM: 608083 MIM: 600745 MIM: 604783
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
APOC2 PubMed Links

Gene Details

Gene
APOC2
Gene Name
apolipoprotein C2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000483.4 229 Missense Mutation AAG,ACG K41T NP_000474.2
Gene
APOC4
Gene Name
apolipoprotein C4
There are no transcripts associated with this gene.

Gene
APOC4-APOC2
Gene Name
APOC4-APOC2 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
CLPTM1
Gene Name
CLPTM1, transmembrane protein
There are no transcripts associated with this gene.

View Full Product Details