Product Details

SNP ID
rs2039205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127613364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGCCGCAGTCTGCAATCTCCCTG[C/G]TTGCGGGGCTCTGCCCTGCCGTGGC
Phenotype
MIM: 602926
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
STXBP1 PubMed Links

Gene Details

Gene
STXBP1
Gene Name
syntaxin binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032221.3 Intron NP_001027392.1
NM_003165.3 Intron NP_003156.1

View Full Product Details