Product Details

SNP ID
rs11552978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:98798724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGCCTCAAGTGGAACACGCAAG[A/G]ATGAGACCAGGCCATATACTGTGAA
Phenotype
MIM: 608698 MIM: 607156
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCBLD2 PubMed Links

Gene Details

Gene
DCBLD2
Gene Name
discoidin, CUB and LCCL domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080927.3 3055 UTR 3 NP_563615.3
XM_011512419.2 3055 UTR 3 XP_011510721.1
Gene
ST3GAL6
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 6
There are no transcripts associated with this gene.

View Full Product Details