Product Details

SNP ID
rs6578
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:194355277 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTATATTATTTATTGTTGATTCT[A/G]TACACCAAATGGATTACAAGCAGCA
Phenotype
MIM: 603104
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPN2 PubMed Links

Gene Details

Gene
CPN2
Gene Name
carboxypeptidase N subunit 2
There are no transcripts associated with this gene.

Gene
LRRC15
Gene Name
leucine rich repeat containing 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135057.2 5853 UTR 3 NP_001128529.2
NM_130830.4 5853 UTR 3 NP_570843.2

View Full Product Details