Product Details

SNP ID
rs1044521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35699714 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAATGGGCTTCGTGATGGGTTGC[C/G]CCGTGGGCATGGCGGCCGGGGCGCT
Phenotype
MIM: 603485 MIM: 604739
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NFS1 PubMed Links

Gene Details

Gene
NFS1
Gene Name
NFS1 cysteine desulfurase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198989.1 224 Intron NP_001185918.1
NM_021100.4 224 Intron NP_066923.3
Gene
RBM39
Gene Name
RNA binding motif protein 39
There are no transcripts associated with this gene.

Gene
ROMO1
Gene Name
reactive oxygen species modulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080748.2 224 Missense Mutation CCC,GCC P28A NP_542786.1
XM_017027678.1 224 Missense Mutation CCC,GCC P28A XP_016883167.1

View Full Product Details