Product Details

SNP ID
rs2574
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:186149761 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACTTGCAGGGCTGGTAGAAAGAAAG[C/G]GGGATTTCCCTTCAGTCTCAGAAAA
Phenotype
MIM: 601854
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DGKG PubMed Links

Gene Details

Gene
DGKG
Gene Name
diacylglycerol kinase gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080744.1 3125 UTR 3 NP_001074213.1
NM_001080745.1 3125 UTR 3 NP_001074214.1
NM_001346.2 3125 UTR 3 NP_001337.2

View Full Product Details