Product Details

SNP ID
rs907938
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:6424231 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTAAAACAGGGGCGAGTGGAAGC[A/G]GCTTTCCCTAAGGCATGACCCCCAG
Phenotype
MIM: 604392
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
AIPL1 PubMed Links

Gene Details

Gene
AIPL1
Gene Name
aryl hydrocarbon receptor interacting protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033054.2 2407 UTR 3 NP_001028226.1
NM_001033055.2 2407 UTR 3 NP_001028227.1
NM_001285399.2 2407 UTR 3 NP_001272328.1
NM_001285400.2 2407 UTR 3 NP_001272329.1
NM_001285401.2 2407 UTR 3 NP_001272330.1
NM_001285402.1 2407 UTR 3 NP_001272331.1
NM_001285403.2 2407 Intron NP_001272332.1
NM_014336.4 2407 UTR 3 NP_055151.3

View Full Product Details