Product Details

SNP ID
rs12610094
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:9102456 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGTCTTCCTAGGTGAGTCAGTAACC[A/G]CAGAACTAATGTACACCCCCAAACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR1M1 PubMed Links

Gene Details

Gene
OR1M1
Gene Name
olfactory receptor family 1 subfamily M member 1
There are no transcripts associated with this gene.

Gene
OR7G2
Gene Name
olfactory receptor family 7 subfamily G member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005193.1 851 Missense Mutation GCG,GTG A284V NP_001005193.1

View Full Product Details