Product Details

SNP ID
rs882020
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:44139144 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATCTGTGCTTTTTGCCTTAGCTTTT[C/T]AACCCTAGACCCCAGACTCTGGGTT
Phenotype
MIM: 138079 MIM: 613993
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GCK PubMed Links

Gene Details

Gene
GCK
Gene Name
glucokinase
There are no transcripts associated with this gene.

Gene
MYL7
Gene Name
myosin light chain 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021223.2 Intron NP_067046.1
XM_005249817.4 Intron XP_005249874.1
XM_011515463.2 Intron XP_011513765.1
XM_011515464.2 Intron XP_011513766.1
XM_011515465.2 Intron XP_011513767.1
XM_017012478.1 Intron XP_016867967.1
XM_017012479.1 Intron XP_016867968.1

View Full Product Details