Product Details

SNP ID
rs14542
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:6650772 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CACCATGAAAAAAGCAGGAAGGGAA[C/T]GGAGAAGAAAAGTGTTGATCACACC
Phenotype
MIM: 608352 MIM: 608524
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ACRBP PubMed Links

Gene Details

Gene
ACRBP
Gene Name
acrosin binding protein
There are no transcripts associated with this gene.

Gene
ING4
Gene Name
inhibitor of growth family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127582.1 1211 UTR 3 NP_001121054.1
NM_001127583.1 1211 UTR 3 NP_001121055.1
NM_001127584.1 1211 UTR 3 NP_001121056.1
NM_001127585.1 1211 UTR 3 NP_001121057.1
NM_001127586.1 1211 UTR 3 NP_001121058.1
NM_016162.3 1211 UTR 3 NP_057246.2
XM_005253698.3 1211 UTR 3 XP_005253755.1
XM_011520964.2 1211 UTR 3 XP_011519266.1
XM_011520965.2 1211 UTR 3 XP_011519267.1
XM_017019392.1 1211 UTR 3 XP_016874881.1
XM_017019393.1 1211 UTR 3 XP_016874882.1

View Full Product Details