Product Details
- SNP ID
-
rs1049303
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:75204170 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCAGGAGCTGCGGGCTCAGTTGGC[A/G]GCCAGGATCTTCTGCACCCAGGGTA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CTRB2
PubMed Links
Gene Details
- Gene
- CTRB2
- Gene Name
- chymotrypsinogen B2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001025200.3 |
816 |
Silent Mutation |
GCC,GCT |
A261A |
NP_001020371.3 |
View Full Product Details