Product Details
- SNP ID
-
hCV11940770
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:85754578 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCACGCCGCCGCCGCCGCCGCCTC[C/A]TGCGCCCCAGAGCCAGGCACCTGGG
- Phenotype
-
- Polymorphism
- C/A, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ATOH8
PubMed Links
Gene Details
- Gene
- ATOH8
- Gene Name
- atonal bHLH transcription factor 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032827.6 |
793 |
Missense Mutation |
CAT,CCT |
H130P |
NP_116216.2 |
View Full Product Details