Product Details

SNP ID
rs3401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:16302247 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTCCCTGAGAAAGAGGTGTGAAA[A/G]GTTCCGCAGAGTCCCTTTAGGGGAG
Phenotype
MIM: 601556 MIM: 139265
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATXN1 PubMed Links

Gene Details

Gene
ATXN1
Gene Name
ataxin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000332.3 7501 UTR 3 NP_000323.2
NM_001128164.1 7501 UTR 3 NP_001121636.1
Gene
GMPR
Gene Name
guanosine monophosphate reductase
There are no transcripts associated with this gene.

View Full Product Details