Product Details
- SNP ID
-
hCV12009031
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:6430137 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCTGGGAGCTGGGGAGGGCAGAG[A/G]CTTGAGCTGCCCCAGGACACAGGGC
- Phenotype
-
MIM: 606351
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ESPN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113421613] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ESPN
- Gene Name
- espin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_031475.2 |
|
Intron |
|
|
NP_113663.2 |
- Gene
- MIR4252
- Gene Name
- microRNA 4252
There are no transcripts associated with this gene.
View Full Product Details