Product Details
- SNP ID
-
rs7925499
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:119216941 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATTATTTATATACTTAAGCAAGT[C/T]GTTGACTTTTCAGCATGTCATCATG
- Phenotype
-
MIM: 165360
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CBL
PubMed Links
Gene Details
- Gene
- CBL
- Gene Name
- Cbl proto-oncogene
View Full Product Details