Product Details

SNP ID
rs4936467
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:119215919 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATTAGCAAATTTTGTGCTGGAAGAA[A/T]ATAGGTAGGAGTAGGCACCTGGAAG
Phenotype
MIM: 165360
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CBL PubMed Links

Gene Details

Gene
CBL
Gene Name
Cbl proto-oncogene
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005188.3 Intron NP_005179.2
XM_011543057.2 Intron XP_011541359.1
XM_017018511.1 Intron XP_016874000.1
Gene
CCDC153
Gene Name
coiled-coil domain containing 153
There are no transcripts associated with this gene.

View Full Product Details