Product Details

SNP ID
hCV12118088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:112425344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTATCTGTCTCATGTAGAATGGA[A/C]CTCTGAAGATCATCGACCGTAAAAA
Phenotype
MIM: 605677
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACSL5 PubMed Links

Gene Details

Gene
ACSL5
Gene Name
acyl-CoA synthetase long-chain family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016234.3 1741 Missense Mutation ACT,CCT T590P NP_057318.2
NM_203379.1 1741 Missense Mutation ACT,CCT T534P NP_976313.1
NM_203380.1 1741 Missense Mutation ACT,CCT T534P NP_976314.1
Gene
ZDHHC6
Gene Name
zinc finger DHHC-type containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303134.1 1741 Intron NP_001290063.1
NM_022494.2 1741 Intron NP_071939.1

View Full Product Details