Product Details

SNP ID
rs2033399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:116429122 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTCTCCAAAATCTGAAACATTTT[A/G]AGCACTAACATGATGCCACAAGCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PNLIPRP3 PubMed Links

Gene Details

Gene
PNLIPRP3
Gene Name
pancreatic lipase related protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011709.2 Intron NP_001011709.2
XM_011539276.1 Intron XP_011537578.1
XM_011539277.1 Intron XP_011537579.1
XM_011539278.1 Intron XP_011537580.1
XM_011539279.1 Intron XP_011537581.1

View Full Product Details