Product Details
- SNP ID
-
rs2449335
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:22548210 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCATAGGAGGCCCTTTGAAGCCCCA[A/C]AATGGGTCATTTGCACTCCCTTCTT
- Phenotype
-
MIM: 114107
MIM: 610795
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PPP3CC
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112961484] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PPP3CC
- Gene Name
- protein phosphatase 3 catalytic subunit gamma
There are no transcripts associated with this gene.
- Gene
- SORBS3
- Gene Name
- sorbin and SH3 domain containing 3
View Full Product Details