Product Details
- SNP ID
-
rs2257906
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:17791486 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGCTCATATTCTGGAAATATGCTAC[C/T]ATTGATATCAAATGCAACATATCCC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C21orf91
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2257907] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C21orf91
- Gene Name
- chromosome 21 open reading frame 91
- Gene
- C21orf91-OT1
- Gene Name
- C21orf91 overlapping transcript 1
There are no transcripts associated with this gene.
View Full Product Details