Product Details
- SNP ID
-
rs2069348
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:163440249 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTCAAGAAATTTGCTTCAACAGAC[C/T]AATTAAGCTCTTTAGACATTTCACA
- Phenotype
-
MIM: 601578
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCNG1
PubMed Links
Gene Details
- Gene
- CCNG1
- Gene Name
- cyclin G1
View Full Product Details