Product Details
- SNP ID
-
rs2069340
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:163437703 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCGGGCCTCTCTCCTCTTGCCTAC[C/G]AGTCCCCTCTCCTCGTAGGCCTCTC
- Phenotype
-
MIM: 601578
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CCNG1
PubMed Links
Gene Details
- Gene
- CCNG1
- Gene Name
- cyclin G1
View Full Product Details