Product Details

SNP ID
rs2071631
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:153547841 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
ATTTGTTGTAGTCACATTCCCGAAA[C/T]TCAGTCTGTGCAAGGGAAGGGTTGG
Phenotype
MIM: 176992 MIM: 114210 MIM: 176991
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928034 PubMed Links

Gene Details

Gene
LOC101928034
Gene Name
uncharacterized LOC101928034
There are no transcripts associated with this gene.

Gene
S100A3
Gene Name
S100 calcium binding protein A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002960.1 230 Silent Mutation GAA,GAG E49E NP_002951.1
Gene
S100A4
Gene Name
S100 calcium binding protein A4
There are no transcripts associated with this gene.

Gene
S100A5
Gene Name
S100 calcium binding protein A5
There are no transcripts associated with this gene.

View Full Product Details