Product Details

SNP ID
rs2240641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:139529552 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGAAACTGTGATGAAGCTGCC[A/G]CCAAGGACAGCAGAGCATGAACAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLEC2L PubMed Links

Gene Details

Gene
CLEC2L
Gene Name
C-type lectin domain family 2 member L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080511.2 Intron NP_001073980.2
XM_017011770.1 Intron XP_016867259.1

View Full Product Details