Product Details
- SNP ID
-
rs2950862
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:23096169 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTGAGACCCTCAATTAGGTGAAAT[A/G]ACTCTTTGTTTGGGCCCTCTTCTCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF730
PubMed Links
Gene Details
- Gene
- ZNF730
- Gene Name
- zinc finger protein 730
View Full Product Details