Product Details

SNP ID
rs2269704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30689176 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGAAGGGCCACGAGGGAGAAGCA[A/G]GGAGACAGTAGAAGAGCATGGGAGG
Phenotype
MIM: 610990
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NRM PubMed Links

Gene Details

Gene
NRM
Gene Name
nurim (nuclear envelope membrane protein)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270707.1 Intron NP_001257636.1
NM_001270708.1 Intron NP_001257637.1
NM_001270709.1 Intron NP_001257638.1
NM_001270710.1 Intron NP_001257639.1
NM_007243.2 Intron NP_009174.1
XM_017010224.1 Intron XP_016865713.1
XM_017010225.1 Intron XP_016865714.1
Gene
PPP1R18
Gene Name
protein phosphatase 1 regulatory subunit 18
There are no transcripts associated with this gene.

View Full Product Details