Product Details

SNP ID
rs2282265
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:95093521 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAAATATTTCTGATCCATTTAAAAA[A/G]GGAAGGATAAGCTACACTGGGCTTT
Phenotype
MIM: 606241
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DICER1 PubMed Links

Gene Details

Gene
DICER1
Gene Name
dicer 1, ribonuclease III
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195573.1 Intron NP_001182502.1
NM_001271282.2 Intron NP_001258211.1
NM_001291628.1 Intron NP_001278557.1
NM_030621.4 Intron NP_085124.2
NM_177438.2 Intron NP_803187.1
XM_011536599.1 Intron XP_011534901.1
XM_011536600.2 Intron XP_011534902.1
XM_011536601.2 Intron XP_011534903.1
XM_011536602.2 Intron XP_011534904.1
XM_011536604.2 Intron XP_011534906.1
XM_011536605.2 Intron XP_011534907.1
XM_017021120.1 Intron XP_016876609.1
XM_017021121.1 Intron XP_016876610.1
XM_017021122.1 Intron XP_016876611.1
XM_017021123.1 Intron XP_016876612.1

View Full Product Details