Product Details

SNP ID
rs2535219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:116499945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCATCTCAAGGTGAAGCCCGGGG[C/T]GGGCGGGCCCAAGTCCCCGCTGAGG
Phenotype
MIM: 601048
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAV2 PubMed Links

Gene Details

Gene
CAV2
Gene Name
caveolin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206747.1 Intron NP_001193676.1
NM_001206748.1 Intron NP_001193677.1
NM_001233.4 Intron NP_001224.1
NM_198212.2 Intron NP_937855.1

View Full Product Details