Product Details

SNP ID
rs2535242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29672897 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGTTGGGCGGCTCTGTGAGGTGG[C/G]CCTTCTGGTGTCTGGAGAGATAGGA
Phenotype
MIM: 159465 MIM: 612192
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MOG PubMed Links

Gene Details

Gene
MOG
Gene Name
myelin oligodendrocyte glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008228.2 1180 Intron NP_001008229.1
NM_001008229.2 1180 Intron NP_001008230.1
NM_001170418.1 1180 Intron NP_001163889.1
NM_002433.4 1180 Intron NP_002424.3
NM_206809.3 1180 Intron NP_996532.2
NM_206810.3 1180 Intron NP_996533.2
NM_206811.3 1180 Intron NP_996534.2
NM_206812.3 1180 Intron NP_996535.2
NM_206814.5 1180 Intron NP_996537.3
XM_005249131.3 1180 Intron XP_005249188.1
Gene
ZFP57
Gene Name
ZFP57 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109809.2 1180 Missense Mutation GCC,GGC A405G NP_001103279.2
XM_006715087.3 1180 Missense Mutation GCC,GGC A333G XP_006715150.1
XM_011514570.2 1180 Missense Mutation GCC,GGC A405G XP_011512872.1

View Full Product Details