Product Details

SNP ID
rs2597220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:14507981 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATACTAACTTTTGATTATCATGAGT[A/G]ACTCCAAAGATCTATGTTATCTAGA
Phenotype
MIM: 600959 MIM: 602854
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COPB1 PubMed Links

Gene Details

Gene
COPB1
Gene Name
coatomer protein complex subunit beta 1
There are no transcripts associated with this gene.

Gene
PSMA1
Gene Name
proteasome subunit alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143937.1 Intron NP_001137409.1
NM_002786.3 Intron NP_002777.1
NM_148976.2 Intron NP_683877.1

View Full Product Details