Product Details
- SNP ID
-
rs2908231
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:9537016 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCAAAGAAAACTAGGGTGGAATTT[T/C]TCCATGTCAGCAGTACATTCTGATA
- Phenotype
-
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OVOS
PubMed Links
Gene Details
- Gene
- OVOS
- Gene Name
- ovostatin
View Full Product Details