Product Details

SNP ID
rs2232872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50486565 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAAGGGGTGGTGCAGAGGGGGC[A/G]TGTGCTTGAGGGTGGGGTCTTGGGT
Phenotype
MIM: 608682 MIM: 606774
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADM2 PubMed Links

Gene Details

Gene
ADM2
Gene Name
adrenomedullin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253845.1 Intron NP_001240774.1
XM_017028940.1 Intron XP_016884429.1
Gene
MIOX
Gene Name
myo-inositol oxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017584.5 Intron NP_060054.4
XM_005261925.3 Intron XP_005261982.1
XM_011530705.1 Intron XP_011529007.1

View Full Product Details