Product Details

SNP ID
rs2279691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240139054 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCATCTTCAGTCCGGAGTTTATTG[A/C]GCGTGAGACCAGGCCTGACTCCTGC
Phenotype
MIM: 607877
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
COPS9 PubMed Links

Gene Details

Gene
COPS9
Gene Name
COP9 signalosome subunit 9
There are no transcripts associated with this gene.

Gene
OTOS
Gene Name
otospiralin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_148961.3 495 UTR 3 NP_683764.1
XM_017003409.1 495 UTR 3 XP_016858898.1
XM_017003410.1 495 UTR 3 XP_016858899.1

View Full Product Details