Product Details

SNP ID
rs2280441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:80948328 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCATTTACTCCACACTCTTGGTCA[A/G]ATTTCTTCACCTCTCAGAACTCTTT
Phenotype
MIM: 608366 MIM: 607783
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEMIP PubMed Links

Gene Details

Gene
CEMIP
Gene Name
cell migration inducing hyaluronan binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293298.1 Intron NP_001280227.1
NM_001293304.1 Intron NP_001280233.1
NM_018689.2 Intron NP_061159.1
Gene
MESDC2
Gene Name
mesoderm development candidate 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015154.2 Intron NP_055969.1

View Full Product Details