Product Details
- SNP ID
-
rs3000858
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
58
- Location
-
Chr.1:12717317 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGGTCCTAGGTTTTCCTACCCTTGA[C/T]GCTCAGATTCTGTAGTCCAGGCTCC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AADACL3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80049576] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AADACL3
- Gene Name
- arylacetamide deacetylase like 3
View Full Product Details