Product Details

SNP ID
rs2305047
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:105107769 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCCGTCGTCGCCGCACGGGCTGG[C/T]TGGGGCTGTGTCTGTGGGAGGCGCC
Phenotype
MIM: 615748
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA1033 PubMed Links

Gene Details

Gene
KIAA1033
Gene Name
KIAA1033
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293640.1 56 UTR 5 NP_001280569.1
NM_015275.2 56 UTR 5 NP_056090.1
XM_011538073.2 56 UTR 5 XP_011536375.1
XM_011538074.2 56 UTR 5 XP_011536376.1
XM_011538077.2 56 UTR 5 XP_011536379.1
XM_017019082.1 56 UTR 5 XP_016874571.1
Gene
LOC414300
Gene Name
uncharacterized LOC414300
There are no transcripts associated with this gene.

View Full Product Details