Product Details

SNP ID
rs2688235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:8448819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCACTTGCCATTGTCCCCTCCAG[C/T]GGGGCCATGCTGCTGCCGCGCCCTC
Phenotype
MIM: 603402 MIM: 614309
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACOX3 PubMed Links

Gene Details

Gene
ACOX3
Gene Name
acyl-CoA oxidase 3, pristanoyl
There are no transcripts associated with this gene.

Gene
TRMT44
Gene Name
tRNA methyltransferase 44 homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152544.2 Intron NP_689757.2
XM_005247950.3 Intron XP_005248007.1
XM_011513406.2 Intron XP_011511708.1
XM_011513407.2 Intron XP_011511709.1
XM_011513408.1 Intron XP_011511710.1
XM_011513409.2 Intron XP_011511711.1
XM_011513410.2 Intron XP_011511712.1
XM_017007811.1 Intron XP_016863300.1

View Full Product Details