Product Details

SNP ID
rs2722899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:28320391 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAGCAAGTGCCAGGAGCACAGGG[C/T]GGCCAGAGCGCTGGCTTCCTCCAAA
Phenotype
MIM: 601459
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PNOC PubMed Links

Gene Details

Gene
PNOC
Gene Name
prepronociceptin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284244.1 Intron NP_001271173.1
NM_006228.4 Intron NP_006219.1
XM_005273532.2 Intron XP_005273589.1
XM_011544559.2 Intron XP_011542861.1
XM_017013578.1 Intron XP_016869067.1

View Full Product Details