Product Details

SNP ID
rs2732903
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:134988968 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTCTTAATTTAAATAGTTTTCCC[A/G]GGTTTATCAAGAATCTTTAAACTTT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
AGBL3 PubMed Links

Gene Details

Gene
AGBL3
Gene Name
ATP/GTP binding protein like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178563.3 Intron NP_848658.3
XM_011516122.2 Intron XP_011514424.1
XM_017012136.1 Intron XP_016867625.1
XM_017012137.1 Intron XP_016867626.1
XM_017012138.1 Intron XP_016867627.1
XM_017012139.1 Intron XP_016867628.1
XM_017012140.1 Intron XP_016867629.1
XM_017012141.1 Intron XP_016867630.1
XM_017012142.1 Intron XP_016867631.1
XM_017012143.1 Intron XP_016867632.1

View Full Product Details